Learn More
MSC E.R. Wagner Rigid Top Plate Caster, 6 inch Polyurethane Wheel, 450 lb Capacity
Supplier:  MSC 57077828
Encompass
This rigid top plate caster features a 6 in diameter, 1-1/2 in wide polyurethane wheel, offering a 450 lb capacity and a 7-3/8 in overall height. Constructed with hardened-heat treated materials and a zinc finish, it provides durability. The caster includes a ball bearing and plate mount, with double row swivel construction, and is corrosion-resistant. It is suitable for food service and general industrial/commercial applications.
  • 6 in wheel diameter
  • Polyurethane wheel material
  • 1-1/2 in wheel width
  • 450 lb load capacity
  • 7-3/8 in overall height
  • Ball bearing type
  • Hardened-heat treated construction
  • Zinc finish
  • Gray and blue wheel color options
  • Plate mount type
  • Rigid style
  • NSF type
  • Double row swivel construction
  • Corrosion-resistant
  • Suitable for food service and general industrial/commercial applications
Catalog No. 50-005-31761