Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Phenolic, 6 inch Wheel Diameter, 450 lb Capacity
Supplier:  MSC 57078206
Encompass
This top plate mount rigid caster features a durable phenolic wheel with a 6 inch diameter and 1-1/2 inch width. Designed for medium-duty applications, it offers a robust 450 lb load capacity. Its hardened-heat treated construction and zinc finish ensure longevity, making it suitable for various industrial settings.
  • Phenolic wheel material
  • 6 inch wheel diameter
  • 1-1/2 inch wheel width
  • 7-3/8 inch overall height
  • 450 lb load capacity
  • Roller bearing type
  • Hardened-heat treated construction
  • Zinc finish
  • Corrosion-resistant
  • Medium-duty applications
Catalog No. 50-005-31812