Learn More
Abcam 6-Thio-GTP (aqueous solution) (Thiogtp), Vac-1-Rac signaling inhibitor, 150UL
Supplier:  Abcam AB146746150UL
Encompass_Preferred
MW 539.24 g/mol, Purity >95%. Vac-1-Rac signaling inhibitor. Azathioprine (ab143064) metabolite. 6-mercaptopurine (ab142389) derivative. Inhibits TNF-α-induced downstream signaling via JNK. Reduces c-Jun, transcription factor-2 and NF-κB activation. Shows immunosuppressant effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4501