Learn More
Medchemexpress LLC HY-128671 10mg Medchemexpress, 6-​Thioinosine CAS:574-25-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12867110MG
Encompass_Preferred
Medchemexpress, HY-128671 10mg 6-​Thioinosine CAS:574-25-4 6-​Thioinosine (6TI) is a purine antimetabolite , acts as an anti-adipogenesis agent, downregulates mRNA levels of PPAR γ and C/EBPα , as well as PPAR γ target protein such as LPL, CD36, aP2, and LXRα [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2626