Learn More
AdipoGen 6-TRITC
SDP
Supplier:  AdipoGen CDXT0035M100
Encompass_Preferred
Chemical. CAS 80724-20-5. Formula C25H21N3O3S. MW 443.52. Synthetic. Useful in the fluorescent labeling of proteins. Labeling efficiency with bovine albumin > 50%.
Catalog No. 50-256-0038