Learn More
Cayman Chemical 6-trns LeukotrIen B4 100ug
SDP
Supplier:  Cayman Chemical 35250100 UG
Encompass_Preferred
A non-enzymatic hydrolysis product of LTA4 with reduced activity compared to LTB4; also produced by oxidative decomposition of CysLTs such as LTC4 in the presence of myeloperoxidase and hypochlorous acid
Catalog No. 50-291-3509