Learn More
Cole-Parmer Cole-Parmer 6-watt UV Lamp with 365nm Wavelength and White Light Tubes, 115VAC
Supplier:  Cole-Parmer 9762010
Encompass_Preferred
Lightweight lamp lets you easily switch from one UV light source to another. A built-in reflector enhances UV illumination, while the durable, extruded metal housing features scratch-resistant powder coating.
Catalog No. NC1230512