Learn More
CELLTREAT Scientific Products 6 Well Tissue Culture Plate, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229105
Encompass_Preferred
CELLTREAT Scientific Products 6 Well Tissue Culture Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with cell growth area of 9.60cm¿. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 50-607-507