Learn More
Chemglass Life Sciences 6-Well Cell Culture Plate, Treated, Flat, Sterile
Supplier: Chemglass Life Sciences CGE3503006
• Made from high clarity, 100% virgin polystyrene• Sterilized by E-beam• Vacuum Plasma tissue culture treatment• Clear lot number for batch traceability• Peel-to-open, individually wrapped• Non-pyrogenic, DNase- and RNase-free