Learn More
Ace Glass, Inc. Beaker, 600mL, jacketed, glass hose connections for 3/8intubing, 152mm inside height, 81mm inside diameter
SDP
Supplier:  Ace Glass, Inc. 534015
Encompass_Preferred
600mL jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 152mm I.H. x 81mm I.D. Beaker does not have a pouring spout.
Catalog No. 50-955-7024