Learn More
Ace Glass, Inc. 600mL jacketed pressure reactor, complete with 28/15 ballpoint I/O, 60mm Ace-Thred four neck head, 50mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
SDP
Supplier:  Ace Glass, Inc. 6386110
Encompass
600mL jacketed pressure reactor, complete with 28/15 ballpoint I/O, 60mm Ace-Thred four neck head, 50mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
Catalog No. 50-972-806