Learn More
Bellco Glass, Inc. 600µL Pipette, MLA, D-Tipper, Silver
SDP
Supplier:  Bellco Glass, Inc. 451900055
Encompass_Preferred
600µL Pipette, MLA, D-Tipper, SilverSingle-stroke “to deliver” operationBuilt in tip-ejectorGlove guardAvailable in 30 fixed volumes ranging from 5 μL- 1000 μLAlso available as Selectable model for set volumes: 50 μL/100 μL/200 μL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorSilverPrimary MaterialAluminum Alloy & Stainless SteelVolume600μLVistaLab Tip Fit SizeLarge 1250μLUniversal FitNoAccuracy±1.00%Precision1.00%
Catalog No. 50-232-9421