Learn More
Thermo Scientific 6020RS Omni Coulometric Sensor (Single Electrode)
Supplier:  Thermo Scientific 6070.2100
Encompass
For Use With Coulometric Detection | Type 6020RS Omni Coulometric Sensor (Single Electrode) | Detection Mode DC | Detector Type Electrochemical | No. of Channels 1 | Reference Electrode Pd | Working Electrodes PG | For Use With (Equipment) Electrochemical Coulometric Detector
Catalog No. NC1752213