Learn More
Triforest Enterprises 60ML sterile PETG square media bottle with tamper evident cap
Supplier:  Triforest Enterprises BGC0060S
Encompass_Preferred
TriForest Square PETG media bottles are ideal for storage of sterile reagents. Bottles are pre-sterilized, SAL 10-6, and include a tamper evident cap. Each tray is packaged for sterile production with multi-barrier packaging.
Catalog No. NC1859845