Learn More
ANALYTICAL SCIENTIFIC INSTRUMENTS US INC 610-NANO-01 1-20NL/MIN
SDP
Supplier:  ANALYTICAL SCIENTIFIC INSTRUMENTS US INC 610NANO01
Encompass
Nano Flow Splitters, Inlet Flow (mL/min.): 250 uL/min., Split Flow (mL/min.): 1.0 - 20 nL/min.
Catalog No. NC1832576