Learn More
ABclonal Technology 610 Rabbit anti-Human CD89
SDP
Supplier:  ABclonal Technology A26636200T
Encompass_Preferred
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils monocytes macrophages and eosinophils where it mediates immunologic responses to pathogens It interacts with IgA-opsonized targets and triggers several immunologic defense processes including phagocytosis antibody-dependent cell-mediated cytotoxicity and stimulation of the release of inflammatory mediators Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene
Catalog No. 50-000-11932