Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 64-Place
Supplier:  Cole-Parmer 0675583/DUP
Encompass_Preferred
Create customized storage box configurations with these freezer boxes. A snug-fitting lid prevents accidental separation if dropped. The boxes can be used in mechanical freezers or liquid nitrogen tanks up to 196C. They are crafted of sturdy cardboard with moisture-repellent coating and designed with small holes on the bottom to allow for drainage. To customize storage needs, cardboard freezer boxes can be purchased separately.
Catalog No. NC1387857
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product