Learn More
ABclonal Technology 647 anti-Human CD26/DPP4
SDP
Supplier:  ABclonal Technology A24411500T
Encompass_Preferred
The protein encoded by this gene is identical to adenosine deaminase complexing protein-2 and to the T-cell activation antigen CD26 It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides
Catalog No. 50-000-09162