Learn More
ABclonal Technology 647 anti-Human CD85d/e/i/j
SDP
Supplier:  ABclonal Technology A27608500T
Encompass_Preferred
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family which is found in a gene cluster at chromosomal region 19q13 4 The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains a transmembrane domain and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13309