Learn More
ABclonal Technology 647 Rabbit anti-Human AXL
SDP
Supplier:  ABclonal Technology A22503500T
Encompass_Preferred
The protein encoded by this gene is a member of the Tyro3-Axl-Mer (TAM) receptor tyrosine kinase subfamily The encoded protein possesses an extracellular domain which is composed of two immunoglobulin-like motifs at the N-terminal followed by two fibronectin type-III motifs It transduces signals from the extracellular matrix into the cytoplasm by binding to the vitamin K-dependent protein growth arrest-specific 6 (Gas6) This gene may be involved in several cellular functions including growth migration aggregation and anti-inflammation in multiple cell types The encoded protein acts as a host cell receptor for multiple viruses including Marburg Ebola and Lassa viruses and is a candidate receptor for the SARS-CoV2 virus
Catalog No. 50-000-07882