Learn More
ABclonal Technology 647 Rabbit anti-Human CD2
SDP
Supplier:  ABclonal Technology A23117100T
Encompass_Preferred
The protein encoded by this gene is a surface antigen found on all peripheral blood T-cells The encoded protein interacts with LFA3 (CD58) on antigen presenting cells to optimize immune recognition A locus control region (LCR) has been found in the 3 flanking sequence of this gene
Catalog No. 50-000-06341