Learn More
ABclonal Technology 647 Rabbit anti-Human CD314
SDP
Supplier:  ABclonal Technology A26181200T
Encompass_Preferred
Natural killer (NK) cells are lymphocytes capable of lysing tumor and virus-infected cells without prior activation as well as regulating immunity NK cells express several calcium-dependent (C-type) lectins which influence their function The NKG2 gene family located in the NK complex includes genes expressed in NK cells This gene encodes a transmembrane protein with a C-type lectin domain binding ligands like MHC class I chain-related A and B proteins and UL-16 binding proteins Ligand-receptor interactions activate NK and T cells aiding immune system recognition of stressed cells This protein and its ligands are potential therapeutic targets for immune diseases and cancers Read-through transcription occurs between this gene and the upstream KLRC4 family member in the same cluster
Catalog No. 50-000-12848