Learn More
ABclonal Technology 647 Rabbit anti-Human CD51
SDP
Supplier:  ABclonal Technology A22299500T
Encompass_Preferred
The product of this gene belongs to the integrin alpha chain family Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha V subunit This subunit associates with beta 1 beta 3 beta 5 beta 6 and beta 8 subunits The heterodimer consisting of alpha V and beta 3 subunits is also known as the vitronectin receptor This integrin may regulate angiogenesis and cancer progression Alternative splicing results in multiple transcript variants Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes
Catalog No. 50-000-05957