Learn More
ABclonal Technology 647 Rabbit anti-Human CD75
SDP
Supplier:  ABclonal Technology A24684200T
Encompass_Preferred
This gene encodes a member of glycosyltransferase family 29 The encoded protein is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates The protein which is normally found in the Golgi but can be proteolytically processed to a soluble form is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6 CD75 and CD76 This gene has been incorrectly referred to as CD75 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-09899