Learn More
ABclonal Technology 647 Rabbit anti-Human CD84
SDP
Supplier:  ABclonal Technology A24579100T
Encompass_Preferred
This gene encodes a membrane glycoprotein that is a member of the signaling lymphocyte activation molecule (SLAM) family This family forms a subset of the larger CD2 cell-surface receptor Ig superfamily The encoded protein is a homophilic adhesion molecule that is expressed in numerous immune cells types and is involved in regulating receptor-mediated signaling in those cells Alternate splicing results in multiple transcript variants
Catalog No. 50-000-10941