Learn More
ABclonal Technology 647 Rabbit anti-Human DLL4
SDP
Supplier:  ABclonal Technology A25866500T
Encompass_Preferred
This gene is a homolog of the Drosophila delta gene The delta gene family encodes Notch ligands that are characterized by a DSL domain EGF repeats and a transmembrane domain
Catalog No. 50-000-09209