Learn More
ABclonal Technology 647 Rabbit anti-Human/ FOXP3
SDP
Supplier:  ABclonal Technology A27685200T
Encompass_Preferred
The protein encoded by this gene is a member of the forkhead winged-helix family of transcriptional regulators Defects in this gene are the cause of immunodeficiency polyendocrinopathy enteropathy X-linked syndrome (IPEX) also known as X-linked autoimmunity-immunodeficiency syndrome Alternatively spliced transcript variants encoding different isoforms have been identified
Catalog No. 50-000-10080