Learn More
ABclonal Technology 647 Rabbit anti-Human Siglec-8
SDP
Supplier:  ABclonal Technology A22190200T
Encompass_Preferred
Sialic acid-binding immunoglobulin (Ig)-like lectins or SIGLECs (e g CD33 (MIM 159590)) are a family of type 1 transmembrane proteins each having a unique expression pattern mostly in hemopoietic cells SIGLEC8 is a member of the CD33-like subgroup of SIGLECs which are localized to 19q13 3-q13 4 and have 2 conserved cytoplasmic tyrosine-based motifs an immunoreceptor tyrosine-based inhibitory motif or ITIM (see MIM 604964) and a motif homologous to one identified in signaling lymphocyte activation molecule (SLAM MIM 603492) that mediates an association with SLAM-associated protein (SAP MIM 300490) (summarized by Foussias et al 2000 [PubMed 11095983])
Catalog No. 50-000-07288