Learn More
ABclonal Technology 647 Rabbit anti-/Rat CD54
SDP
Supplier:  ABclonal Technology A24895200T
Encompass_Preferred
This gene encodes an integral membrane protein that binds leukocyte adhesion protein LFA-1 It participates in the innate immune response
Catalog No. 50-000-09567