Learn More
MOUSER ELECTRONICS 69-1/2INX36YD TAPE
Supplier:  MOUSER ELECTRONICS 51769TAPE1/2
Encompass
Description/Function: 69 Electrical Tape - OEM Glass Cloth With Silicone Pressure Color: White Material: Acetate Cloth Adhesive Type: Silicone Width - in: 0.5 in Width - mm: 12.7 mm Length - in: 36 yd Length - mm: 32.92 m Brand: 3M Electronic Specialty
Catalog No. NC3449931