Learn More
Cayman Chemical 6aMethylprednIsoLN 21acet 50mg
SDP
Supplier:  Cayman Chemical 3579950 MG
Encompass_Preferred
A synthetic glucocorticoid and an ester form of methylprednisolone; reduces IL-1β-induced production of PGE2 and increases in MMP-13 levels in equine osteochondral and synovial explant cocultures from 1-100 nM; intramuscular injection induces retinal lipid peroxidation and apoptosis in rabbits at 20 mg/kg
Catalog No. 50-291-1405