Learn More
Cayman Chemical 6chLRoMDMAhydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 900194410 MG
Encompass_Preferred
Differs structurally from 3,4-MDMA in that it has a chlorine atom added at the two position of the phenyl ring.
Catalog No. 50-289-4579