Learn More
Aat Bioquest 6-FAM Azide
SDP
Supplier:  Aat Bioquest 133
Encompass_Preferred
6-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
Catalog No. 50-297-9273