Learn More
Aat Bioquest 6-FAM, SE [6-Carboxyfluorescein, succinimidyl ester] [CAS 92557-81-8]
SDP
Supplier:  Aat Bioquest 117
Encompass_Preferred
6-FAM, SE is the amine-reactive succinimidyl ester of single isomer 6-FAM acid.
Catalog No. 50-297-9355