Learn More
Medchemexpress LLC 6h-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-6-one, 2,3,3a,9a-tetrahydro-3-hydroxy-2-(hydroxymethyl)-7-methyl | 22423-26-3 | 240.21 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00849110G
Encompass_Preferred
TK-112690 is an inhibitor of UPP1 (uridine phosphorylase 1). It inhibits murine small intestinal uridine phosphorylase (UPase) with an IC50 of 12.5 μM and human small intestinal UPase with an IC50 of 20.0 μM in vitro. It has been shown to increase plasma uridine concentration in mice and can be utilized for research into cancer and pulmonary fibrosis.
  • Inhibits UPP1 (uridine phosphorylase 1)
  • Increases plasma uridine concentration in mice
  • Utilized for research into cancer and pulmonary fibrosis
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature is acceptable if less than 2 weeks
Catalog No. 50-004-47494