Learn More
Medchemexpress LLC 6H-Pyrido[2,3-b]benzodiazepin-6-one, 5,11-dihydro-11-[2-(4-methyl-1-piperazinyl)acetyl]-, hydrochloride (1:2) | 29868-97-1 | 692100 | 99.8% | 424.32 | 500MG
SDP
Supplier:  Medchemexpress LLC HY17037500MG
Encompass_Preferred
Pirenzepine (LS 519) dihydrochloride is a selective M1 mAChR (muscarinic acetylcholine receptor) antagonist with poor penetration of the blood-brain barrier. It reduces gastric acid secretion and muscle spasm, and can be used in peptic ulcer research. Pirenzepine dihydrochloride also demonstrates anti-proliferative activity against cancer cells.
  • Reduces gastric acid secretion
  • Reduces muscle spasm
  • Used in peptic ulcer research
  • Shows anti-proliferative activity to cancer cells
Catalog No. 50-003-86089