Learn More
Apexbio Technology LLC 6H05 1469338-01-9 5mg
SDP
Supplier:  Apexbio Technology LLC A89025
Encompass_Preferred
6H05 (CAS 1469338-01-9) is a selective allosteric inhibitor targeting the K-ras (G12C) mutant protein It acts by covalently binding to the mutated cysteine residue stabilizing K-ras in an inactive conformation and thereby blocking downstream signaling pathways 6H05 is utilized to investigate the molecular mechanisms underlying K-ras (G12C) mutation-driven oncogenesis and signal transduction It serves as a tool for assessing K-ras (G12C)-mediated tumorigenic processes in cell-based and animal models facilitating early-stage pharmacological evaluation and target validation studies in the context of anti-cancer drug research
Catalog No. 50-000-95932