Learn More
Cayman Chemical 6H05trIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 1798510 MG
Encompass_Preferred
An allosteric inhibitor of oncogenic K-Ras(G12C); disrupts both switch-I and switch-II, altering the native nucleotide preference to favor GDP over GTP, which impedes binding to Raf
Catalog No. 50-296-5954