Learn More
USA Scientific Inc 6pl upright freezer rack
Supplier:  USA Scientific Inc 26610900
Encompass
6-place (6 long x 1 high) upright freezer rack for multiple well plates, 22 1/8 in L x 5 1/2 in W x 9 1/4 in H, stainless steel
Catalog No. 50-274-5120