Learn More
Medchemexpress LLC 6PPD | 793-24-8 | 98.46% | 268.40 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01168950G
Encompass_Preferred
6PPD is a rubber antioxidant that scavenges ozone and forms nitro radicals. Exposure to 6PPD reduces the hatching rate, impairs spontaneous locomotor activity, shortens body length, and causes malformations in zebrafish embryos. 6PPD also induces oxidative stress in zebrafish embryos. It is for research use only and not sold to patients.
  • Rubber antioxidant that scavenges ozone and forms nitro radicals.
Catalog No. 50-004-52865