Learn More
TARGETMOL CHEMICALS INC 6PPD-Q 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T784745MG
Encompass_Preferred
Also available in 1 mg, 10 mg and bulk. Please contact Fisher for quotes. 6PPD-Q (6PPD-Quinone), a contaminant present in the environment and soil, is a derivative of the tire antioxidant N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylenediamine (6PPD), which impairs the quality of spermatozoa in mice and induces impaired male fertility. Purity 98.43%
Catalog No. 50-287-7084