Learn More
Medchemexpress LLC 6PPD-quinone-d5 | 2750119-14-1 | 99.4% | 303.41 | C18H17D5N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY152039S10MG
Encompass_Preferred
6PPD-quinone-d5 is the deuterium-labeled isotopologue of 6PPD-quinone supplied as an analytical standard for laboratory research. It is intended for use as an internal standard in mass spectrometry workflows for environmental, toxicological, and method development applications, and is provided with characterization data to support quantitative analyses.
  • Deuterium-labeled internal standard for quantitative LC/GC-MS.
  • High chemical and isotopic purity for reliable quantitation.
  • Certificate of analysis and safety data available for verification.
  • Supplied as a stable solid appropriate for laboratory storage.
  • Suitable for environmental and toxicology method development.
Catalog No. 50-002-86395