Learn More
Enzo Life Sciences (6R)-5,6,7,8-Tetrahydro-L-biopterin. dihydrochloride (5mg). CAS: 69056-38-8
SDP
Supplier:  Enzo Life Sciences ALX440001M005
Encompass_Preferred
Cofactor of nitric oxide synthases (NOS). Alternative name: (6R)-BH4 . 2 HCl, [6R-[6R*(1R*,2S*)]]-2-Amino-6-(1,2-dihydroxypropyl)-5,6,7,8-tetrahydro-4(1H)-pteridinone . 2 HCl, Sapropterin dihydrochloride. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9264