Learn More
Cayman Chemical 6R5 6 7 8tetrahydroLBIo 5mg
SDP
Supplier:  Cayman Chemical 818805 MG
Encompass_Preferred
A cofactor for production of aromatic amino acids, neurotransmitters, and nitric oxide
Catalog No. 50-289-6594