Learn More
Medchemexpress LLC (6S,9S,12S)-benzyl 12-benzyl-9-isobutyl-2,2-dimethyl-4,7,10-trioxo-6-phenethyl-3-oxa-5,8,11-triazatridecan- | 868540-15-2 | 99.9% | C37H47N3O6 | 5 G

Supplier: Medchemexpress LLC HY798635G
This product is a phenylalanine derivative intended for research use only. Amino acids and their derivatives, like this compound, are recognized as beneficial ergogenic dietary substances due to their influence on anabolic hormones, fuel supply during exercise, mental performance under stress, and prevention of exercise-induced muscle damage.
- Chemical formula: C37H47N3O6
- Molecular weight: 629.79
- CAS number: 868540-15-2
- Appearance: solid
- Color: white to off-white
- Purity: 99.89%
- Classified as amino acid derivatives, peptides, and peptide fragments
- Primarily for research purposes
- May influence anabolic hormones and fuel supply during exercise
- Can enhance mental performance under stress
- Helps prevent exercise-induced muscle damage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.