Learn More
Cayman Chemical 6trns12epI LeukotrIen B4 100ug
SDP
Supplier:  Cayman Chemical 35265100 UG
Encompass_Preferred
A non-enzymatic hydrolysis product of LTA4 with significantly reduced activity compared to LTB4
Catalog No. 50-291-3432