Learn More
Electron Microscopy Sciences Folding Magnifier 6x
SDP
Supplier:  Electron Microscopy Sciences 68112
Encompass_Preferred
Lightweight, compact, and simple; it comes with a measuring scale on the base frame. Base size is 1 square inch opening; marked every 1/8" on two sides and every millimeter on two other sides. Comes with 6X magnifier lens in a black anodized aluminum body. Folds flat when it is not in use.
Catalog No. 50-259-81