Learn More
Alkali Scientific 6x29mm PP Insert Conical Base with Polyspring and Graduation, 100/Pk
Supplier: Alkali Scientific CV5505
The Alkali Scientific™ 6 × 29 mm PP Insert is designed for precise sample handling and reliable analytical performance. Made from durable polypropylene (PP), this insert features a conical base to maximize sample recovery and reduce residual volume. The integrated polyspring ensures consistent positioning within standard autosampler vials, while the graduated markings allow accurate volume measurement. Ideal for chromatography and other high precision laboratory applications, these inserts provide excellent chemical resistance and durability. Supplied in packs of 100, they offer consistent performance and ease of use for high throughput workflows. Key Features 6 × 29 mm for standard autosampler vial compatibility Made from durable polypropylene (PP) with excellent chemical resistance Conical base design maximizes sample recovery and minimizes residual volume Integrated polyspring ensures consistent positioning within vials Graduated markings allow accurate volume measuremen