Learn More
Sklar Instruments Scissors, Lister Bandage Scissors, Sklar, 7-1/2 in., Blunt/Blunt, Bandage Scissors, Angled, Smooth, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 111072LFT
Encompass_Preferred
Scissors, Lister Bandage Scissors, Sklar, 7-1/2 in., Blunt/Blunt, Bandage Scissors, Angled, Smooth, Stainless Steel, EA. Sklar Left-Handed Lister Bandage Scissors are a versatile instrument with multiple uses, forged specifically for the left-handed user. Lister Bandage Scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without cutting the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. This product is angled with smooth, blunt/blunt tips and a length of 7-1/4 inches.
Catalog No. 50-109-3307