Learn More
Medchemexpress LLC 7-[(2,5-dioxopyrrolidin-1-yl)methyl]-4-(4-fluorophenyl)quinoline-2-carboxamide | 1432728-49-8 | 99.3% | C21H16FN3O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13355510MG
Encompass_Preferred
mGluR2 antagonist 1 is a potent, selective negative allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2), reported with an IC50 of 9 nM. It is orally bioavailable, exhibits excellent brain permeability, and is supplied as a high-purity solid for preclinical and in vitro research applications.
  • Potent mGluR2 negative allosteric modulator (IC50 = 9 nM).
  • Orally bioavailable and brain permeable.
  • High purity suitable for research use.
  • Provided as a solid in multiple package sizes for experimental flexibility.
  • Intended for neuroscience and pharmacology research applications.
Catalog No. 50-002-58451